missing translation for 'onlineSavingsMsg'
Learn More

Memantine Hydrochloride, TRC

Product Code. 30134216
Change view
Click to view available options
Quantity:
50 mg
100 mg
1 g
Unit Size:
100mg
1g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134216 1 g 1g
30012055 100 mg 100mg
30012459 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134216 Supplier Toronto Research Chemicals Supplier No. TRCM2180001G
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 41100-52-1
Molecular Formula C12 H21 N . Cl H
Molecular Weight (g/mol) 215.76
Synonym Memantine Hydrochloride, Tricyclo[3.3.1.13,7]decan-1-amine, 3,5-dimethyl-, hydrochloride (1:1)1-Adamantanamine, 3,5-dimethyl-, hydrochloride (7CI), Tricyclo[3.3.1.13,7]decan-1-amine, 3,5-dimethyl-, hydrochloride (9CI), 1-Amino-3,5-dimethyladamantane hydrochloride, 3,5-Dimethyl-1-aminoadamantane hydrochloride, 3,5-Dimethyladamantan-1-amine hydrochloride, 3,5-Dimethyltricyclo[3.3.1.13,7]decan-1-amine hydrochloride, Akatinol, Axura, Ebix, Ebixia, Ebixza, Mantine, Memantine hydrochloride, Memary, NSC 102290, Namenda, SUN Y7017, from Axura
IUPAC Name 3,5-dimethyladamantan-1-amine;hydrochloride
SMILES Cl.CC12CC3CC(C)(C1)CC(N)(C3)C2

Specifications

Quantity 1 g
InChI Formula InChI=1S/C12H21N.ClH/c1-10-3-9-4-11(2,6-10)8-12(13,5-9)7-10;/h9H,3-8,13H2,1-2H3;1H
Formula Weight 215.1441
Percent Purity >95%
Chemical Name or Material Memantine Hydrochloride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.