accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 454678-91-2

IUPAC Name: [4-(1,3-thiazol-2-yl)phenyl]methanol
Synonyms: [4-(1,3-thiazol-2-yl)phenyl]methanol
Molecular Weight (g/mol): 191.25
Molecular Formula: C10H9NOS
InChi Key: QOLUYHARONOGNO-UHFFFAOYSA-N
SMILES: OCC1=CC=C(C=C1)C1=NC=CS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results