accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CNHONH3CO

CAS RN 50-12-4

IUPAC Name: 5-ethyl-3-methyl-5-phenylimidazolidine-2,4-dione
Synonyms: mephenytoin
Molecular Weight (g/mol): 218.26
Molecular Formula: C12H14N2O2
InChi Key: GMHKMTDVRCWUDX-UHFFFAOYNA-N
SMILES: CCC1(NC(=O)N(C)C1=O)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)
Certifications/Compliance 
  • (1)
  • (1)
Industry Type 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results