missing translation for 'onlineSavingsMsg'
Learn More

rac-Mephenytoin, TRC

Product Code. 30135628
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135628 5 mg 5mg
30135086 10 mg 10mg
17952725 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135628 Supplier Toronto Research Chemicals Supplier No. TRCM2249855MG
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-12-4
Molecular Formula C12 H14 N2 O2
Molecular Weight (g/mol) 218.25
Synonym Mephenytoin, 2,4-Imidazolidinedione, 5-ethyl-3-methyl-5-phenyl-Hydantoin, 5-ethyl-3-methyl-5-phenyl- (6CI,7CI,8CI), (±)-5-Ethyl-3-methyl-5-phenylhydantoin, (±)-Mephenytoin, (±)-Mesantoin, 3-Ethylnirvanol, 3-Methyl-5-ethyl-5-phenyl-2,4-imidazolidinedione, 3-Methyl-5-ethyl-5-phenylhydantoin, 5-Ethyl-3-methyl-5-phenyl-2,4(3H,5H)-imidazoledione, 5-Ethyl-3-methyl-5-phenylhydantoin, 5-Ethyl-3-methyl-5-phenylimidazolidin-2,4-dione, DL-Mephenytoin, Epiazin, Epilan, Fenantoin, Gerot-Epilan, Insulton, Mephenytoin, Mesantoin, Mesdontoin, Mesontoin, Methoin, Methylphenetoin, Metydan, NSC 34652, Phenantoin, Phenylethylmethylhydantoin, Sacerno, Sedantional, Sedantoin, Triantoin
IUPAC Name 5-ethyl-3-methyl-5-phenylimidazolidine-2,4-dione
SMILES CCC1(NC(=O)N(C)C1=O)c2ccccc2

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C12H14N2O2/c1-3-12(9-7-5-4-6-8-9)10(15)14(2)11(16)13-12/h4-8H,3H2,1-2H3,(H,13,16)
Formula Weight 218.1055
Chemical Name or Material Mephenytoin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.