accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3SSH3CNN

CAS RN 5147-80-8

IUPAC Name: 2-[bis(methylsulfanyl)methylidene]propanedinitrile
Synonyms: 2-[bis(methylsulfanyl)methylidene]propanedinitrile
Molecular Weight (g/mol): 170.25
Molecular Formula: C6H6N2S2
InChi Key: FICQFRCPSFCFBY-UHFFFAOYSA-N
SMILES: CSC(SC)=C(C#N)C#N
Molecular Weight (g/mol) 
  • (5)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (5)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)