missing translation for 'onlineSavingsMsg'
Learn More

2-[Di(methylthio)methylidene]malononitrile, TRC

Product Code. 30388232
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30388232 1 g
30371501 5 g
30383790 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30388232 Supplier Toronto Research Chemicals Supplier No. TRCD4620801G
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5147-80-8
Molecular Formula C6H6N2S2
Molecular Weight (g/mol) 170.25
IUPAC Name 2-[bis(methylsulfanyl)methylidene]propanedinitrile
SMILES CSC(=C(C#N)C#N)SC

Specifications

Quantity 1 g
InChI Formula InChI=1S/C6H6N2S2/c1-9-6(10-2)5(3-7)4-8/h1-2H3
Formula Weight 169.9972
Chemical Name or Material 2-[Di(methylthio)methylidene]malononitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.