accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COFOCH3

CAS RN 52189-63-6

IUPAC Name: 1-fluoro-3,5-dimethoxybenzene
Synonyms: 1-fluoro-3,5-dimethoxybenzene
Molecular Weight (g/mol): 156.16
Molecular Formula: C8H9FO2
InChi Key: IWFKMNAEFPEIOY-UHFFFAOYSA-N
SMILES: COC1=CC(OC)=CC(F)=C1
Boiling Point 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results