missing translation for 'onlineSavingsMsg'
Learn More

1-Fluoro-3,5-dimethoxybenzene, 98%

Product Code. 11456566
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11456566 100 g 100g
11436566 5 g 5g
11446566 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11456566 Supplier Thermo Scientific Alfa Aesar Supplier No. B24810.22
 Restricted Item
Estimated Shipment: 09-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 52189-63-6 | C8H9FO2 | 156.16 g/mol

1-Fluoro-3,5-dimethoxybenzene is used in the preparation of 5-fluoro-benzene-1,3-diol using reagents like tetrabutylammnoium iodide and boron trichloirde.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Fluoro-3,5-dimethoxybenzene is used in the preparation of 5-fluoro-benzene-1,3-diol using reagents like tetrabutylammnoium iodide and boron trichloirde.

Solubility
Slightly miscible with water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52189-63-6
Molecular Formula C8H9FO2
Molecular Weight (g/mol) 156.16
MDL Number MFCD00012445
InChI Key IWFKMNAEFPEIOY-UHFFFAOYSA-N
Synonym 1,3-dimethoxy-5-fluorobenzene, benzene, 1-fluoro-3,5-dimethoxy, 3,5-dimethoxyfluorobenzene, pubchem3049, acmc-209wz0, 3,5-dimethoxy fluorobenzene, ksc595k6b, 3,5-dimethoxy-1-fluorobenzene, 1-fluoro-3,5-dimethoxy benzene, benzene,1-fluoro-3,5-dimethoxy
PubChem CID 2774257
IUPAC Name 1-fluoro-3,5-dimethoxybenzene
SMILES COC1=CC(OC)=CC(F)=C1

Specifications

Density 1.17
Boiling Point 86°C to 88°C (7 mmHg)
Flash Point 91°C (196°F)
Refractive Index 1.5025
Quantity 100 g
Solubility Information Slightly miscible with water.
Formula Weight 156.16
Percent Purity 98%
Chemical Name or Material 1-Fluoro-3,5-dimethoxybenzene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.