accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ONONCl

CAS RN 53062-98-9

IUPAC Name: 4-(2-chloropyridine-3-carbonyl)morpholine
Synonyms: 4-(2-chloropyridine-3-carbonyl)morpholine
Molecular Weight (g/mol): 226.66
Molecular Formula: C10H11ClN2O2
InChi Key: SPSWDQBZCCVHNI-UHFFFAOYSA-N
SMILES: ClC1=C(C=CC=N1)C(=O)N1CCOCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results