accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CONNOO

CAS RN 5446-92-4

IUPAC Name: 2-methoxy-5-nitropyridine
Synonyms: 2-methoxy-5-nitropyridine
Molecular Weight (g/mol): 154.13
Molecular Formula: C6H6N2O3
InChi Key: WUPLOZFIOAEYMG-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=N1)[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results