CAS RN 55-55-0
4-Methylaminophenol sulfate, 99%, extra pure
CAS: 55-55-0 Molecular Formula: C14H18N2O2·H2SO4 Molecular Weight (g/mol): 344.38 MDL Number: MFCD00013140 Synonym: Metol
4-Methylaminophenol sulfate, ACS reagent, 99.0-101.5%
CAS: 55-55-0 Molecular Formula: C14H18N2O2·H2SO4 Molecular Weight (g/mol): 344.38 Synonym: Metol
4-(Methylamino)phenol sulfate, 99%
CAS: 55-55-0 Molecular Formula: C14H18N2O2·H2SO4 MDL Number: MFCD00013140 Synonym: Bis(4-hydroxy-N-methylanilinium) sulfate; Metol
4-(Methylamino)phenol sulfate, ACS, 99.0-101.5%
CAS: 55-55-0 MDL Number: MFCD00013140 Synonym: Bis(4-hydroxy-n-methylanilinium) sulfate; Metol
Metol, TRC
CAS: 55-55-0 Molecular Formula: 2(C7H9NO) • H2SO4 Molecular Weight (g/mol): 2123.169807 Synonym: 4-(Methylamino)-phenol Sulfate (2:1) (salt),p-(Methylamino)-phenol Sulfate (2:1) (salt),4-(Methylamino)phenol Sulfate (2:1) Salt,Armol’ Elon,Elon (developer),Genol,Graphol,Metatyl,Metol,N-Methyl-4-hydroxyaniline hemisulfate,N-Methyl-p-aminophenol sulfate,N-Methyl-p-aminophenol sulfuric acid hemisalt,N-Methyl-p-aminophenol sulfuric acid salt (2:1),Photol,Pictol,Planetol,Rhodol,p-Methylaminophenol Sulfate,p-Methylaminophenol Sulphate (2:1) SMILES: OC1=CC=C(NC)C=C1.OC1=CC=C(NC)C=C1.[H2SO4]