missing translation for 'onlineSavingsMsg'
Learn More

Metol, TRC

Product Code. 30474326
Change view
Click to view available options
Quantity:
5 g
10 g
100 g
Unit Size:
100g
10g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30474326 100 g 100g
30467310 10 g 10g
30470393 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30474326 Supplier Toronto Research Chemicals Supplier No. TRCM338720100G
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 55-55-0
Molecular Formula 2(C7H9NO) • H2SO4
Molecular Weight (g/mol) 2123.169807
Synonym 4-(Methylamino)-phenol Sulfate (2:1) (salt), p-(Methylamino)-phenol Sulfate (2:1) (salt), 4-(Methylamino)phenol Sulfate (2:1) Salt, Armol’ Elon, Elon (developer), Genol, Graphol, Metatyl, Metol, N-Methyl-4-hydroxyaniline hemisulfate, N-Methyl-p-aminophenol sulfate, N-Methyl-p-aminophenol sulfuric acid hemisalt, N-Methyl-p-aminophenol sulfuric acid salt (2:1), Photol, Pictol, Planetol, Rhodol, p-Methylaminophenol Sulfate, p-Methylaminophenol Sulphate (2:1)
SMILES OC1=CC=C(NC)C=C1.OC1=CC=C(NC)C=C1.[H2SO4]

Specifications

Quantity 100 g
InChI Formula InChI=1S/2C7H9NO.H2O4S/c2*1-8-6-2-4-7(9)5-3-6;1-5(2,3)4/h2*2-5,8-9H,1H3;(H2,1,2,3,4)
Chemical Name or Material Metol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.