accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3ONOCH3

CAS RN 57597-62-3

IUPAC Name: 3,3-dimethoxypropanenitrile
Synonyms: 3,3-dimethoxypropanenitrile
Molecular Weight (g/mol): 115.13
Molecular Formula: C5H9NO2
InChi Key: JYHSJQNYYLGMEI-UHFFFAOYSA-N
SMILES: COC(CC#N)OC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results