accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NH2ClF

CAS RN 57946-56-2

IUPAC Name: 4-chloro-2-fluoroaniline
Synonyms: 4-chloro-2-fluoroaniline
Molecular Weight (g/mol): 145.56
Molecular Formula: C6H5ClFN
InChi Key: CSFDTBRRIBJILD-UHFFFAOYSA-N
SMILES: NC1=CC=C(Cl)C=C1F
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results