CAS RN 58-58-2
Puromycin dihydrochloride, 98%
CAS: 58-58-2 Molecular Formula: C22H31Cl2N7O5 Molecular Weight (g/mol): 544.43 MDL Number: MFCD00150080 InChI Key: MKSVFGKWZLUTTO-MLYJQVMKNA-N PubChem CID: 131632508 SMILES: Cl.Cl.COC1=CC=C(C[C@H](N)C(=O)N[C@@H]2[C@@H](CO)O[C@H]([C@@H]2O)N2C=NC3=C(N=CN=C23)N(C)C)C=C1
Thermo Scientific Chemicals Puromycin dihydrochloride hydrate, 99%
CAS: 58-58-2 Molecular Formula: C22H31Cl2N7O5 Molecular Weight (g/mol): 544.43 MDL Number: MFCD00150080 InChI Key: MKSVFGKWZLUTTO-MLYJQVMKNA-N Synonym: puromycin dihydrochloride PubChem CID: 131632508 IUPAC Name: (2S)-2-amino-N-[(2S,3S,4S,5R)-5-[6-(dimethylamino)purin-9-yl]-4-hydroxy-2-(hydroxymethyl)oxolan-3-yl]-3-(4-methoxyphenyl)propanamide;dihydrochloride SMILES: Cl.Cl.COC1=CC=C(C[C@H](N)C(=O)N[C@@H]2[C@@H](CO)O[C@H]([C@@H]2O)N2C=NC3=C(N=CN=C23)N(C)C)C=C1
Puromycin dihydrochloride, 10 mg/ml in distilled water, sterile-filtered
CAS: 58-58-2 Molecular Formula: C22H31Cl2N7O5 Molecular Weight (g/mol): 544.43 MDL Number: MFCD00150080 InChI Key: MKSVFGKWZLUTTO-MLYJQVMKNA-N PubChem CID: 439530 ChEBI: CHEBI:17939 SMILES: Cl.Cl.COC1=CC=C(C[C@H](N)C(=O)N[C@@H]2[C@@H](CO)O[C@H]([C@@H]2O)N2C=NC3=C(N=CN=C23)N(C)C)C=C1
Puromycin Dihydrochloride, Fisher BioReagents™
Inhibits protein synthesis | CAS: 58-58-2 | C22H31Cl2N7O5 | 544.43 g/mol
Puromycin Dihydrochloride,, MP Biomedicals
CAS: 58-58-2 Molecular Formula: C22H31Cl2N7O5 Molecular Weight (g/mol): 544.43 MDL Number: MFCD00150080 InChI Key: MKSVFGKWZLUTTO-MLYJQVMKNA-N Synonym: Stylomycin,6-dimethylamino-9-(3'-deoxy-3'-(p-methoxy-L-phenylalanyamino)- B-D-ribofuranosyl)-purine PubChem CID: 131632508 SMILES: Cl.Cl.COC1=CC=C(C[C@H](N)C(=O)N[C@@H]2[C@@H](CO)O[C@H]([C@@H]2O)N2C=NC3=C(N=CN=C23)N(C)C)C=C1
Melford Puromycin Dihydrochloride
Puromycin Dihydrochloride is a potent antibiotic.
Puromycin Dihydrochloride, TRC
CAS: 58-58-2 Molecular Formula: C22 H29 N7 O5 . 2 Cl H Molecular Weight (g/mol): 544.43 Synonym: 3'-[[(2S)-2-amino-3-(4-methoxyphenyl)-1-oxopropyl]amino]-3'-deoxy-N,N-dimethyl-adenosine hydrochloride (1:2),Puromycin dihydrochloride,Stylomycin dihydrochloride IUPAC Name: (2S)-2-amino-N-[(2S,3S,4R,5R)-5-[6-(dimethylamino)purin-9-yl]-4-hydroxy-2-(hydroxymethyl)oxolan-3-yl]-3-(4-methoxyphenyl)propanamide;dihydrochloride SMILES: Cl.Cl.COc1ccc(C[C@H](N)C(=O)N[C@@H]2[C@@H](CO)O[C@H]([C@@H]2O)n3cnc4c(ncnc34)N(C)C)cc1
TOKU-E™ Puromycin Dihydrochloride ReadyMade™ Solution
Puromycin Dihydrochloride ReadyMade™ Solution is provided as a sterile-filtered solution of Puromycin Dihydrochloride in 20 mM HEPES buffer.