accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H2NNONHNONH(S)OHOOHONNH

CAS RN 59-30-3

IUPAC Name: (2S)-2-[(4-{[(2-amino-4-oxo-1,4-dihydropteridin-6-yl)methyl]amino}phenyl)formamido]pentanedioic acid
Synonyms: folic acid apo-folic foliamin PGA dosfolat B activ pteroylglutamic acid vitamin B9 pteroyl-L-glutamic acid vitamin bc vitamin be
Molecular Weight (g/mol): 441.4
Molecular Formula: C19H19N7O6
InChi Key: OVBPIULPVIDEAO-LBPRGKRZSA-N
SMILES: NC1=NC(=O)C2=NC(CNC3=CC=C(C=C3)C(=O)N[C@@H](CCC(O)=O)C(O)=O)=CN=C2N1
Molecular Weight (g/mol) 
  • (9)
  • (2)
  • (14)
Quantity 
  • (1)
  • (3)
  • (6)
  • (6)
  • (1)
  • (4)
  • (1)
Physical Form 
  • (2)
  • (3)
  • (8)
  • (4)
Percent Purity 
  • (1)
  • (3)
  • (4)
  • (3)
Certifications/Compliance 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (4)
  • (1)
  • (1)
  • (7)
  • (3)
  • (5)
  • (3)
  • (1)
  • (9)
  • (2)
  • (14)
  • (1)
  • (3)
  • (6)
  • (6)
  • (1)
  • (4)
  • (1)
  • (2)
  • (3)
  • (8)
  • (4)
  • (1)
  • (3)
  • (4)
  • (3)
  • (2)
  • (1)
  • (3)
  • (1)