accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OHONH2(S)NHNHClOHH

CAS RN 5934-29-2

IUPAC Name: hydrogen (2S)-2-amino-3-(1H-imidazol-5-yl)propanoic acid hydrate chloride
Synonyms: hydrogen L-histidine hydrate chloride
Molecular Weight (g/mol): 209.63
Molecular Formula: C6H12ClN3O3
InChi Key: CMXXUDSWGMGYLZ-XRIGFGBMSA-N
SMILES: [H+].O.[Cl-].N[C@@H](CC1=CN=CN1)C(O)=O
Quantity 
  • (5)
  • (2)
  • (4)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (4)
Percent Purity 
  • (2)
  • (4)
  • (3)
  • (4)
Grade 
  • (2)
  • (4)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (4)
  • (2)
  • (4)
  • (7)
  • (2)
  • (3)
  • (5)
  • (2)
  • (4)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (4)
  • (2)
  • (4)
  • (3)
  • (4)
  • (2)
  • (4)
  • (1)
  • (1)