missing translation for 'onlineSavingsMsg'
Learn More

L-Histidine monohydrochloride monohydrate, Cell Culture Reagent

Product Code. 15410197
Change view
Click to view available options
Quantity:
10 g
100 g
500 g
1 kg
Unit Size:
100g
10g
1kg
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15410197 100 g 100g
15400197 10 g 10g
15420197 500 g 500g
15430197 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15410197 Supplier Thermo Scientific Alfa Aesar Supplier No. J61465.22
  • ONLINE EXCLUSIVE PRICE
    113.22€
    92.65€ / 100g
    Save 20.57€
    18% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 5934-29-2 | C6H12ClN3O3 | 209.63 g/mol

L-Histidine is used in cell culture media formulations used in biomanufacturing. L-Histidine has been used to study cultures of the human T-lymphoblastic leukemia cell line MOLT-4 to study modulation of apoptosis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Histidine is used in cell culture media formulations used in biomanufacturing. L-Histidine has been used to study cultures of the human T-lymphoblastic leukemia cell line MOLT-4 to study modulation of apoptosis.

Solubility
Freely soluble in water

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5934-29-2
Molecular Formula C6H12ClN3O3
Molecular Weight (g/mol) 209.63
MDL Number MFCD00151027
InChI Key CMXXUDSWGMGYLZ-XRIGFGBMSA-N
Synonym l-histidine hydrochloride hydrate, l-histidine hydrochloride monohydrate, s-2-amino-3-1h-imidazol-4-yl propanoic acid hydrochloride hydrate, h-his-oh.hcl.h2o, l-histidine monohydrochloride monohydrate, l-histidine, monohydrochloride, monohydrate, l-histidin hydrate hydrochloride, histidine, monohydrochloride, monohydrate, l, h-his-ohhclh2o, l-histidine, hydrochloride, monohydrate
PubChem CID 165377
IUPAC Name (2S)-2-amino-3-(1H-imidazol-4-yl)propanoic acid hydrate hydrochloride
SMILES O.Cl.N[C@@H](CC1=CNC=N1)C(O)=O

Specifications

Melting Point ∽240°C (decomposition)
Color White
Beilstein 4168261
Merck Index 14,4720
Solubility Information Freely soluble in water
Quantity 100 g
Formula Weight 209.63 (191.62 Anhydrous)
Optical Rotation +10° (c=5 in 5N HCl)
Grade Cell Culture Reagent
Physical Form Powder
Chemical Name or Material L-Histidine monohydrochloride monohydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.