accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 599-79-1

HOOONHNSOONHN(Z)

CAS RN 599-79-1

IUPAC Name: (3Z)-6-oxo-3-(2-{4-[(pyridin-2-yl)sulfamoyl]phenyl}hydrazin-1-ylidene)cyclohexa-1,4-diene-1-carboxylic acid
Synonyms: sulfasalazine
Molecular Weight (g/mol): 398.39
Molecular Formula: C18H14N4O5S
InChi Key: OQANPHBRHBJGNZ-BKUYFWCQSA-N
SMILES: OC(=O)C1=C\C(C=CC1=O)=N/NC1=CC=C(C=C1)S(=O)(=O)NC1=CC=CC=N1
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results