missing translation for 'onlineSavingsMsg'
Learn More

Sulfasalazine, TRC

Product Code. 30137097
Change view
Click to view available options
Quantity:
50 mg
5 g
10 g
Unit Size:
10g
50mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137097 10 g 10g
30136332 5 g 5g
18092220 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137097 Supplier Toronto Research Chemicals Supplier No. TRCS69908410G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 599-79-1
Molecular Formula C18 H14 N4 O5 S
Molecular Weight (g/mol) 398.39
Synonym Sulfasalazine, 2-Hydroxy-5-[2-[4-(pyridin-2-ylsulfamoyl)phenyl]diazenyl]benzoic acid, Benzoic acid, 2-hydroxy-5-[2-[4-[(2-pyridinylamino)sulfonyl]phenyl]diazenyl]-Benzoic acid, 2-hydroxy-5-[[4-[(2-pyridinylamino)sulfonyl]phenyl]azo]- (9CI), Salicylic acid, 5-[[p-(2-pyridylsulfamoyl)phenyl]azo]- (6CI,7CI,8CI), 2-Hydroxy-5-((4-((2-pyridinylamino)sulfonyl)phenyl)azo)benzoic acid, 5-[4-(2-Pyridylsulfamyl)phenylazo]-2-hydroxybenzoic acid, 5-[p-(2-Pyridylsulfamoyl)phenylazo]salicylic acid, 5-[p-(2-Pyridylsulfamyl)phenylazo]salicylic acid, Azopyrin, Azopyrine, Azulfidine, Azulfidine EN, Azulfin, Benzosulfa, Colo-Pleon, NSC 203730, NSC 667219, Reupirin, Salazopyridin, Salazopyrin, Salazopyrine, Salazosulfapyridin, Salazosulfapyridine, Salicylazosulfapyridine, Salisulf, Sulfasalazin, Sulfasalazine, Sulphasalazine
IUPAC Name 2-hydroxy-5-[(E)-[4-(pyridin-2-ylsulfamoyl)phenyl]diazenyl]benzoic acid
SMILES OC(=O)c1cc(ccc1O)N=Nc2ccc(cc2)S(=O)(=O)Nc3ccccn3

Specifica

Quantity 10 g
InChI Formula InChI=1S/C18H14N4O5S/c23-16-9-6-13(11-15(16)18(24)25)21-20-12-4-7-14(8-5-12)28(26,27)22-17-3-1-2-10-19-17/h1-11,23H,(H,19,22)(H,24,25)/b21-20+
Formula Weight 398.0685
Chemical Name or Material Sulfasalazine
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.