accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 62108-07-0

CH3HOH3CH2C

CAS RN 62108-07-0

IUPAC Name: 4-propylhept-1-en-4-ol
Synonyms: 4-propylhept-1-en-4-ol
Molecular Weight (g/mol): 156.27
Molecular Formula: C10H20O
InChi Key: RLUDGGHEFUDPLJ-UHFFFAOYSA-N
SMILES: CCCC(O)(CCC)CC=C
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results