missing translation for 'onlineSavingsMsg'
Learn More

4-n-Propyl-1-hepten-4-ol, 98%, Thermo Scientific™

Product Code. 11444377
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11444377 100 g 100g
11424377 5 g 5g
11434377 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11444377 Supplier Thermo Scientific Alfa Aesar Supplier No. B20429.22
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62108-07-0
Molecular Formula C10H20O
Molecular Weight (g/mol) 156.269
MDL Number MFCD00049206
InChI Key RLUDGGHEFUDPLJ-UHFFFAOYSA-N
Synonym 4-n-propyl-1-hepten-4-ol, 4-propyl-1-hepten-4-ol, 1-hepten-4-ol, 4-propyl, allyldipropylcarbinol, allyl di-n-propyl carbinol, 4-hydroxy-4-propyl-1-heptene
PubChem CID 551382
IUPAC Name 4-propylhept-1-en-4-ol
SMILES CCCC(CCC)(CC=C)O

Specifications

Quantity 100 g
Formula Weight 156.27
Percent Purity 98%
Chemical Name or Material 4-n-Propyl-1-hepten-4-ol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.