accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 63134-33-8

IUPAC Name: 4-[4-(benzyloxy)benzenesulfonyl]phenol
Synonyms: 4-[4-(benzyloxy)benzenesulfonyl]phenol
Molecular Weight (g/mol): 340.39
Molecular Formula: C19H16O4S
InChi Key: UWPJWCBDMZHMTN-UHFFFAOYSA-N
SMILES: OC1=CC=C(C=C1)S(=O)(=O)C1=CC=C(OCC2=CC=CC=C2)C=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results