accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COOOCl

CAS RN 638-07-3

IUPAC Name: ethyl 4-chloro-3-oxobutanoate
Synonyms: ethyl 4-chloro-3-oxobutanoate
Molecular Weight (g/mol): 164.59
Molecular Formula: C6H9ClO3
InChi Key: OHLRLMWUFVDREV-UHFFFAOYSA-N
SMILES: CCOC(=O)CC(=O)CCl
Molecular Weight (g/mol) 
  • (3)
  • (6)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
Boiling Point 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (6)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)