accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 68890-66-4

H3CONHOCH3H3CCH3H3CNH2HO

CAS RN 68890-66-4

IUPAC Name: 1-hydroxy-4-methyl-6-(2,4,4-trimethylpentyl)-1,2-dihydropyridin-2-one; 2-aminoethan-1-ol
Synonyms: piroctone olamine
Molecular Weight (g/mol): 298.43
Molecular Formula: C16H30N2O3
InChi Key: BTSZTGGZJQFALU-UHFFFAOYNA-N
SMILES: NCCO.CC(CC1=CC(C)=CC(=O)N1O)CC(C)(C)C
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)