accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ClSH2NCl

CAS RN 69622-81-7

IUPAC Name: 2,5-dichlorobenzene-1-carbothioamide
Synonyms: 2,5-dichlorobenzenecarbothioamide
Molecular Weight (g/mol): 206.08
Molecular Formula: C7H5Cl2NS
InChi Key: YQORNGRDGWVQOD-UHFFFAOYSA-N
SMILES: NC(=S)C1=C(Cl)C=CC(Cl)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results