accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OHO

CAS RN 698-27-1

IUPAC Name: 2-hydroxy-4-methylbenzaldehyde
Synonyms: 2-hydroxy-4-methylbenzaldehyde
Molecular Weight (g/mol): 136.15
Molecular Formula: C8H8O2
InChi Key: JODRRPJMQDFCBJ-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=O)C(O)=C1
Quantity 
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (3)
Boiling Point 
  • (3)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (3)
  • (3)
  • (2)