accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 7164-98-9

NN

CAS RN 7164-98-9

IUPAC Name: 1-phenyl-1H-imidazole
Synonyms: 1-phenyl-1H-imidazole
Molecular Weight (g/mol): 144.18
Molecular Formula: C9H8N2
InChi Key: SEULWJSKCVACTH-UHFFFAOYSA-N
SMILES: C1=CN(C=N1)C1=CC=CC=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results