accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 74037-50-6

IUPAC Name: 3-ethoxy-2-nitropyridine
Synonyms: 3-ethoxy-2-nitropyridine
Molecular Weight (g/mol): 168.15
Molecular Formula: C7H8N2O3
InChi Key: WBEVIQNYYGPESH-UHFFFAOYSA-N
SMILES: CCOC1=CC=CN=C1[N+]([O-])=O
Boiling Point 
  • (4)
  • (1)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)