accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 75820-45-0

CH3H3CCH3SOHN

CAS RN 75820-45-0

IUPAC Name: 4-tert-butyl-2,3-dihydro-1,3-thiazol-2-one
Synonyms: 4-tert-butyl-3H-1,3-thiazol-2-one
Molecular Weight (g/mol): 157.23
Molecular Formula: C7H11NOS
InChi Key: DZGLMEMWTFPPTN-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CSC(=O)N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results