CAS RN 773057-50-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(S)-1-Trichloromethyl-1,2,3,4-Tetrahydro-Beta-carboline Hydrochloride, TRC
CAS: 773057-50-4 Molecular Formula: C12H11Cl3N2 . HCl Molecular Weight (g/mol): 326 Synonym: (S)-2,3,4,9-Tetrahydro-1-(trichloromethyl)-1H-pyrido[3,4-b]indole Hydrochloride,(R)-TaClo Hydrochloride IUPAC Name: (1S)-1-(trichloromethyl)-2,3,4,9-tetrahydro-1H-pyrido[3,4-b]indole;hydrochloride SMILES: Cl.ClC(Cl)(Cl)[C@H]1NCCc2c1[nH]c3ccccc23