accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 78266-06-5

H3CH3CHNONOHOOHOCH3Br

CAS RN 78266-06-5

IUPAC Name: 2-({[(3-bromo-2,4,6-trimethylphenyl)carbamoyl]methyl}(carboxymethyl)amino)acetic acid
Synonyms: ({[(3-bromo-2,4,6-trimethylphenyl)carbamoyl]methyl}(carboxymethyl)amino)acetic acid
Molecular Weight (g/mol): 387.23
Molecular Formula: C15H19BrN2O5
InChi Key: MHPZZZZLAQGTHT-UHFFFAOYSA-N
SMILES: CC1=CC(C)=C(NC(=O)CN(CC(O)=O)CC(O)=O)C(C)=C1Br

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results