Learn More
(3-Bromo-2,4,6-trimethylphenylcarbamoyl)methyliminodiacetic acid, 98+%
Product Code. 11392877
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11392877 5 g 5g
11382877 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11392877 Supplier Thermo Scientific Alfa Aesar Supplier No. L16086.06
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
CAS: 78266-06-5 | C15H19BrN2O5 | 387.23 g/mol
(3-Bromo-2,4,6-trimethylphenylcarbamoyl)methyliminodiacetic acid is used in the diagnostic field of medicine and in the assessment of hepatobillary transport.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
(3-Bromo-2,4,6-trimethylphenylcarbamoyl)methyliminodiacetic acid is used in the diagnostic field of medicine and in the assessment of hepatobillary transport.
Solubility
Soluble in dimethylsulphoxide and ethanol. Insoluble in water.
Notes
Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 78266-06-5
Molecular Formula C15H19BrN2O5
Molecular Weight (g/mol) 387.23
MDL Number MFCD00216974
InChI Key MHPZZZZLAQGTHT-UHFFFAOYSA-N
Synonym mebrofenin, choletec, mebrofenine inn-french, mebrofeninum inn-latin, mebrofenino inn-spanish, unii-7pv0b6ed98, 3-bromo-2,4,6-trimethylphenylcarbamoyl methyliminodiacetic acid, 3-bromomesityl carbamoyl methyl imino diacetic acid, n-2-3-bromo-2,4,6-trimethylphenyl amino-2-oxoethyl-n-carboxymethyl glycine, mebrofenine
PubChem CID 54158
IUPAC Name 2-[[2-(3-bromo-2,4,6-trimethylanilino)-2-oxoethyl]-(carboxymethyl)amino]acetic acid
SMILES CC1=CC(=C(C(=C1NC(=O)CN(CC(=O)O)CC(=O)O)C)Br)C
Specifications
Melting Point 192°C to 196°C
Quantity 5 g
Solubility Information Soluble in dimethylsulphoxide and ethanol. Insoluble in water.
Formula Weight 387.24
Percent Purity ≥98%
Chemical Name or Material (3-Bromo-2,4,6-trimethylphenylcarbamoyl)methyliminodiacetic acid