accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

BBrFHOOH

CAS RN 849062-37-9

IUPAC Name: (3-bromo-5-fluorophenyl)boronic acid
Synonyms: 3-bromo-5-fluorophenylboronic acid
Molecular Weight (g/mol): 218.82
Molecular Formula: C6H5BBrFO2
InChi Key: UVKKALLRVWTDOY-UHFFFAOYSA-N
SMILES: OB(O)C1=CC(F)=CC(Br)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results