accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ClOCl

CAS RN 85-29-0

IUPAC Name: (2-chlorophenyl)(4-chlorophenyl)methanone
Synonyms: (2-chlorophenyl)(4-chlorophenyl)methanone
Molecular Weight (g/mol): 251.11
Molecular Formula: C13H8Cl2O
InChi Key: YXMYPHLWXBXNFF-UHFFFAOYSA-N
SMILES: ClC1=CC=C(C=C1)C(=O)C1=CC=CC=C1Cl
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results