Learn More
2,4'-Dichlorobenzophenone, 98%, Thermo Scientific™
Product Code. 11436750
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11436750 25 g 25g
11426750 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11436750 Supplier Thermo Scientific Alfa Aesar Supplier No. L07648.14
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 85-29-0
Molecular Formula C13H8Cl2O
Molecular Weight (g/mol) 251.11
MDL Number MFCD00038744
InChI Key YXMYPHLWXBXNFF-UHFFFAOYSA-N
Synonym 2,4'-dichlorobenzophenone, 2-chlorophenyl 4-chlorophenyl methanone, methanone, 2-chlorophenyl 4-chlorophenyl, 2-chlorophenyl-4-chlorophenyl methanone, unii-h3n0v78i65, benzophenone, 2,4'-dichloro, di2-chlorophenyl ketone, pubchem3373, o,p-dichlorobenzophenone, o,p'-dichlorobenzophenone
PubChem CID 66558
IUPAC Name (2-chlorophenyl)-(4-chlorophenyl)methanone
SMILES ClC1=CC=C(C=C1)C(=O)C1=CC=CC=C1Cl
Specifications
Melting Point 63°C to 66°C
Quantity 25 g
Beilstein 1959090
Formula Weight 251.11
Percent Purity 98%
Chemical Name or Material 2,4'-Dichlorobenzophenone