accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3ONBOHHO

CAS RN 854373-97-0

IUPAC Name: (2-ethoxypyridin-3-yl)boronic acid
Synonyms: 2-ethoxypyridin-3-ylboronic acid
Molecular Weight (g/mol): 166.97
Molecular Formula: C7H10BNO3
InChi Key: FXUMKSCYKPOZOO-UHFFFAOYSA-N
SMILES: CCOC1=NC=CC=C1B(O)O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results