missing translation for 'onlineSavingsMsg'
Learn More

2-Ethoxypyridine-3-boronic acid, 98%

Product Code. 11345679
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11345679 250 mg 250mg
11325679 1 g 1g
11335679 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11345679 Supplier Thermo Scientific Alfa Aesar Supplier No. L20422.MD
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 854373-97-0 | C7H10BNO3 | 166.97 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 854373-97-0
Molecular Formula C7H10BNO3
Molecular Weight (g/mol) 166.97
MDL Number MFCD06201036
InChI Key FXUMKSCYKPOZOO-UHFFFAOYSA-N
Synonym 2-ethoxypyridine-3-boronic acid, 2-ethoxy-3-pyridineboronic acid, 2-ethoxypyridin-3-yl boronic acid, 2-ethoxy-3-pyridyl boronic acid, 3-borono-2-ethoxypyridine, pubchem5119, acmc-209q5z, 2-ethoxypyridine-3-boronicacid, 2-ethoxy-3-pyridinyl boronic acid, 2-ethoxy-pyridin-3-yl-boronic acid
PubChem CID 11309733
IUPAC Name (2-ethoxypyridin-3-yl)boronic acid
SMILES CCOC1=NC=CC=C1B(O)O

Specifications

Quantity 250 mg
Formula Weight 166.97
Percent Purity 98%
Chemical Name or Material 2-Ethoxypyridine-3-boronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.