accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NOOOH

CAS RN 86649-57-2

IUPAC Name: 2-(pyridin-3-yloxy)acetic acid
Synonyms: (pyridin-3-yloxy)acetic acid
Molecular Weight (g/mol): 153.14
Molecular Formula: C7H7NO3
InChi Key: MBEPWLCDXKKANL-UHFFFAOYSA-N
SMILES: OC(=O)COC1=CN=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results