accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 883536-06-9

CH3OFOF

CAS RN 883536-06-9

IUPAC Name: 4-ethoxy-3,5-difluorobenzaldehyde
Synonyms: 4-ethoxy-3,5-difluorobenzaldehyde
Molecular Weight (g/mol): 186.16
Molecular Formula: C9H8F2O2
InChi Key: IOKIEIKNSQIONF-UHFFFAOYSA-N
SMILES: CCOC1=C(F)C=C(C=O)C=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results