accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COOFFFOCH3

CAS RN 886500-08-9

IUPAC Name: 1-[2-methoxy-4-(trifluoromethoxy)phenyl]ethan-1-one
Synonyms: 1-[2-methoxy-4-(trifluoromethoxy)phenyl]ethanone
Molecular Weight (g/mol): 234.17
Molecular Formula: C10H9F3O3
InChi Key: YAMJXZPRMMJVTQ-UHFFFAOYSA-N
SMILES: COC1=C(C=CC(OC(F)(F)F)=C1)C(C)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results