accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ONOHNSHNHNOClClClF

CAS RN 905973-89-9

IUPAC Name: 2,2-diphenyl-N-(2,2,2-trichloro-1-{[(4-fluoro-3-nitrophenyl)carbamothioyl]amino}ethyl)acetamide
Synonyms: 2,2-diphenyl-N-(2,2,2-trichloro-1-{[(4-fluoro-3-nitrophenyl)carbamothioyl]amino}ethyl)acetamide
Molecular Weight (g/mol): 555.83
Molecular Formula: C23H18Cl3FN4O3S
InChi Key: HLCDNLNLQNYZTK-UHFFFAOYNA-N
SMILES: [O-][N+](=O)C1=CC(NC(=S)NC(NC(=O)C(C2=CC=CC=C2)C2=CC=CC=C2)C(Cl)(Cl)Cl)=CC=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results