CAS RN 923942-34-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Methyl 2-[3-Cyano-4-(2-methylpropoxy)phenyl]-4-methylthiazole-5-carboxylate, TRC
CAS: 923942-34-1 Molecular Formula: C17H18N2O3S Molecular Weight (g/mol): 330.4 Synonym: Methyl 2-[3-Cyano-4-(2-methylpropoxy)phenyl]-4-methyl-5-thiazolecarboxylic Acid Ester,Methyl 2-(3-Cyano-4-isobutoxyphenyl)-4-methylthiazole-5-carboxylate Acetate; IUPAC Name: methyl 2-[3-cyano-4-(2-methylpropoxy)phenyl]-4-methyl-1,3-thiazole-5-carboxylate SMILES: CC1=C(C(OC)=O)SC(C2=CC(C#N)=C(OCC(C)C)C=C2)=N1