accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 931-97-5

HON

CAS RN 931-97-5

IUPAC Name: 1-hydroxycyclohexane-1-carbonitrile
Synonyms: 1-hydroxycyclohexane-1-carbonitrile
Molecular Weight (g/mol): 125.17
Molecular Formula: C7H11NO
InChi Key: ZDBRPNZOTCHLSP-UHFFFAOYSA-N
SMILES: OC1(CCCCC1)C#N
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results