accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 937-14-4

OHOOCl

CAS RN 937-14-4

IUPAC Name: 3-chlorobenzene-1-carboperoxoic acid
Synonyms: mcpba m-cpba m-chlorobenzoyl hydroperoxide
Molecular Weight (g/mol): 172.56
Molecular Formula: C7H5ClO3
InChi Key: NHQDETIJWKXCTC-UHFFFAOYSA-N
SMILES: OOC(=O)C1=CC=CC(Cl)=C1
Molecular Weight (g/mol) 
  • (7)
  • (3)
Quantity 
  • (1)
  • (3)
  • (3)
  • (1)
  • (2)
Percent Purity 
  • (3)
  • (4)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (3)
  • (7)
  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (2)
  • (3)
  • (4)
  • (4)