missing translation for 'onlineSavingsMsg'
Learn More

3-Chloroperoxybenzoic acid, 50-55%, cont. ca 10% 3-chlorobenzoic acid, balance water

Product Code. 11359973
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 11359973

Brand: Thermo Scientific Alfa Aesar L00286.36

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

CAS: 937-14-4 | C7H5ClO3 | 172.56 g/mol

3-Chloroperoxybenzoic acid is a strong oxidizing agent used in the oxidation reactions such as aldehydes and ketones to esters (Bayer-Villiger-Oxidation), olefines to epoxides, sulfides to sulfoxides and sulfones, and amines to nitroalkanes, N-oxides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Chloroperoxybenzoic acid is a strong oxidizing agent used in the oxidation reactions such as aldehydes and ketones to esters (Bayer-Villiger-Oxidation), olefines to epoxides, sulfides to sulfoxides and sulfones, and amines to nitroalkanes, N-oxides.

Solubility
Soluble in potassium hydroxide, chloroform and acetone. Insoluble in water.

Notes
Heat sensitive. Incompatible with reactive with reducing agents, combustible materials, organic materials and alkalies.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 937-14-4
Molecular Formula C7H5ClO3
Molecular Weight (g/mol) 172.56
MDL Number MFCD00002127
InChI Key NHQDETIJWKXCTC-UHFFFAOYSA-N
Synonym 3-chloroperoxybenzoic acid, 3-chloroperbenzoic acid, mcpba, m-chloroperbenzoic acid, m-chloroperoxybenzoic acid, meta-chloroperoxybenzoic acid, benzenecarboperoxoic acid, 3-chloro, m-chlorobenzoyl hydroperoxide, meta-chloroperbenzoic acid, 3-chlorobenzoperoxoic acid
PubChem CID 70297
ChEBI CHEBI:52091
IUPAC Name 3-chlorobenzenecarboperoxoic acid
SMILES OOC(=O)C1=CC=CC(Cl)=C1

Specifications

Melting Point 64°C to 66°C
Quantity 500 g
UN Number UN3106
Beilstein 608317
Solubility Information Soluble in potassium hydroxide,chloroform and acetone. Insoluble in water.
Formula Weight 172.57
Percent Purity 50 to 55%
Assay cont. ≈10% 3-chlorobenzoic acid, balance water
Chemical Name or Material 3-Chloroperoxybenzoic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.