accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 96-17-3

CH3H3CO

CAS RN 96-17-3

IUPAC Name: 2-methylbutanal
Synonyms: 2-methylbutanal α-2-methyl-n-butanal
Molecular Weight (g/mol): 86.13
Molecular Formula: C5H10O
InChi Key: BYGQBDHUGHBGMD-UHFFFAOYNA-N
SMILES: CCC(C)C=O
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)