accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OOOOHCH3CH3CH3CH3CH3H3CH3CH3CH3COHOOOCH3H3CCH3H3C

CAS RN 97600-49-2

IUPAC Name: ethyl 2-{[5,11,17,23-tetra-tert-butyl-27-(2-ethoxy-2-oxoethoxy)-26,28-dihydroxypentacyclo[19.3.1.1³,⁷.1⁹,¹³.1¹⁵,¹⁹]octacosa-1(25),3,5,7(28),9(27),10,12,15(26),16,18,21,23-dodecaen-25-yl]oxy}acetate
Synonyms: ethyl 2-{[5,11,17,23-tetra-tert-butyl-27-(2-ethoxy-2-oxoethoxy)-26,28-dihydroxypentacyclo[19.3.1.1³,⁷.1⁹,¹³.1¹⁵,¹⁹]octacosa-1(25),3,5,7(28),9(27),10,12,15(26),16,18,21,23-dodecaen-25-yl]oxy}acetate
Molecular Weight (g/mol): 821.11
Molecular Formula: C52H68O8
InChi Key: RCVLXPWJVXHCMD-UHFFFAOYSA-N
SMILES: CCOC(=O)COC1=C2CC3=CC(=CC(CC4=C(OCC(=O)OCC)C(CC5=C(O)C(CC1=CC(=C2)C(C)(C)C)=CC(=C5)C(C)(C)C)=CC(=C4)C(C)(C)C)=C3O)C(C)(C)C
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results