accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CSNClO

CAS RN 97936-43-1

IUPAC Name: 2-(methylsulfanyl)pyridine-3-carbonyl chloride
Synonyms: 2-(methylsulfanyl)pyridine-3-carbonyl chloride
Molecular Weight (g/mol): 187.64
Molecular Formula: C7H6ClNOS
InChi Key: OCEMBWMMHUSVMT-UHFFFAOYSA-N
SMILES: CSC1=NC=CC=C1C(Cl)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results