accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OHSOHOO

CAS RN 98-67-9

IUPAC Name: 4-hydroxybenzene-1-sulfonic acid
Synonyms: phenolsulfonic acid sulfocarbolic acid hydroxybenzenesulfonic acid hydroxybenzene-4-sulfonic acid
Molecular Weight (g/mol): 174.17
Molecular Formula: C6H6O4S
InChi Key: FEPBITJSIHRMRT-UHFFFAOYSA-N
SMILES: OC1=CC=C(C=C1)S(O)(=O)=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results